Assay Details
Gene Symbol: GTF3C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650931.1 | XP_023506699.1 | 288 | 101 | ||
XM_023650932.1 | XP_023506700.1 | 288 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
