Assay Details
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504045.6 | XP_001504095.2 | 1590 | 82 | ||
XM_005596881.3 | XP_005596938.1 | 1608 | 82 | |||
XM_005596882.3 | XP_005596939.1 | 1760 | 82 | |||
XM_014734583.2 | XP_014590069.1 | 1601 | 82 | |||
XM_023650940.1 | XP_023506708.1 | 1366 | 82 | |||
XM_023650942.1 | XP_023506710.1 | 656 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
