Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917929.5 | XP_001917964.1 | 498 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
