Assay Details
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610331.3 | XP_005610388.1 | 206 | 69 | ||
XM_014740109.2 | XP_014595595.1 | 214 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
