Assay Details
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610341.3 | XP_005610398.3 | 2726 | 146 | ||
XM_014740181.2 | XP_014595667.1 | 1202 | 146 | |||
XM_023641432.1 | XP_023497200.1 | 2718 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
