Assay Details
Gene Symbol: MYBPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610369.3 | XP_005610426.1 | 3006 | 61 | ||
XM_014740116.2 | XP_014595602.1 | 8466 | 61 | |||
XM_014740117.2 | XP_014595603.1 | 8466 | 61 | |||
XM_014740118.2 | XP_014595604.1 | 8466 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
