Assay Details
Gene Symbol: SARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494097.6 | XP_001494147.1 | 1184 | 61 | ||
XM_005610372.3 | XP_005610429.1 | 1185 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
