Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493959.5 | XP_001494009.1 | 479 | 56 | ||
XM_005610374.3 | XP_005610431.1 | 481 | 56 | |||
XM_014740093.2 | XP_014595579.1 | 482 | 56 | |||
XM_014740094.2 | XP_014595580.1 | 480 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
