Assay Details
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493837.5 | XP_001493887.1 | 205 | 81 | ||
GenBank mRNA | JL615997.1 | - | 2 - 3 | 136 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
