Assay Details
Gene Symbol: FAM102B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641518.1 | XP_023497286.1 | 758 | 79 | ||
XM_023641519.1 | XP_023497287.1 | 756 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
