Assay Details
Gene Symbol: GFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641644.1 | XP_023497412.1 | 333 | 78 | ||
XM_023641645.1 | XP_023497413.1 | 875 | 78 | |||
XM_023641646.1 | XP_023497414.1 | 322 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
