Assay Details
Gene Symbol: ZUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504142.6 | XP_001504192.1 | 237 | 103 | ||
XM_023650967.1 | XP_023506735.1 | 238 | 103 | |||
XM_023650968.1 | XP_023506736.1 | 238 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504142.6, XM_023650967.1, XM_023650968.1 transcripts.
Set Membership: |
![]() ![]() |
