Assay Details
Gene Symbol: ZUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504142.6 | XP_001504192.1 | 1215 | 83 | ||
XM_023650967.1 | XP_023506735.1 | 1216 | 83 | |||
XM_023650968.1 | XP_023506736.1 | 925 | 83 |
