Assay Details
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650991.1 | XP_023506759.1 | 1799 | 72 | ||
XM_023650992.1 | XP_023506760.1 | 2046 | 72 | |||
XM_023650993.1 | XP_023506761.1 | 1892 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
