Assay Details
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504213.5 | XP_001504263.1 | 2012 | 65 | ||
XM_023651009.1 | XP_023506777.1 | 2064 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
