Assay Details
Gene Symbol: CENPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362370.4 | XP_003362418.1 | 298 | 85 | ||
XM_023651032.1 | XP_023506800.1 | 302 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
