Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596925.3 | XP_005596982.1 | 379 | 95 | ||
XM_023651085.1 | XP_023506853.1 | 319 | 95 | |||
XM_023651086.1 | XP_023506854.1 | 319 | 95 | |||
XM_023651087.1 | XP_023506855.1 | 361 | 95 | |||
GenBank mRNA | JL620430.1 | - | 1 - 2 | 56 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
