Assay Details
Gene Symbol: FN3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490214.6 | XP_001490264.1 | 592 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
