Assay Details
Gene Symbol: HEXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490394.6 | XP_001490444.3 | 301 | 84 | ||
XM_014735780.2 | XP_014591266.1 | 301 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001490394.6, XM_014735780.2 transcripts.
Set Membership: |
![]() ![]() |
