Assay Details
Gene Symbol: HEXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490394.6 | XP_001490444.3 | 504 | 55 | ||
XM_014735780.2 | XP_014591266.1 | 504 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
