Assay Details
Gene Symbol: ASPSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737643.2 | XP_014593129.1 | 771 | 64 | ||
XM_023651750.1 | XP_023507518.1 | 427 | 64 | |||
XM_023651751.1 | XP_023507519.1 | 427 | 64 | |||
XM_023651752.1 | XP_023507520.1 | 427 | 64 | |||
XM_023651753.1 | XP_023507521.1 | 427 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
