Assay Details
Gene Symbol: MCRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488427.5 | XP_001488477.1 | 188 | 111 | ||
XM_005597054.3 | XP_005597111.1 | 238 | 111 | |||
XM_005597056.3 | XP_005597113.1 | 538 | 111 | |||
XM_014737752.2 | XP_014593238.1 | 566 | 111 | |||
XM_023651772.1 | XP_023507540.1 | 519 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
