Assay Details
Gene Symbol: MRPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488514.4 | XP_001488564.1 | 1004 | 83 | ||
XM_005597061.3 | XP_005597118.1 | 1039 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
