Assay Details
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651865.1 | XP_023507633.1 | 1272 | 66 | ||
GenBank mRNA | JL620713.1 | - | 10 - 11 | 1014 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
