Assay Details
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651882.1 | XP_023507650.1 | 558 | 90 | ||
XM_023651883.1 | XP_023507651.1 | 560 | 90 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651882.1 | XP_023507650.1 | 558 | 90 | ||
XM_023651883.1 | XP_023507651.1 | 560 | 90 |