Assay Details
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651882.1 | XP_023507650.1 | 1182 | 68 | ||
XM_023651883.1 | XP_023507651.1 | 1184 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
