Assay Details
Gene Symbol: CANT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490865.4 | XP_001490915.1 | 788 | 76 | ||
XM_005597091.3 | XP_005597148.1 | 763 | 76 | |||
XM_014738216.2 | XP_014593702.2 | 674 | 76 | |||
XM_014738225.2 | XP_014593711.1 | 1197 | 76 | |||
XM_023651896.1 | XP_023507664.1 | 1102 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
