Assay Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492039.6 | XP_001492089.3 | 1842 | 63 | ||
XM_005597150.3 | XP_005597207.1 | 1624 | 63 | |||
XM_005597151.3 | XP_005597208.1 | 1989 | 63 | |||
XM_005597152.3 | XP_005597209.1 | 2472 | 63 | |||
XM_005597153.3 | XP_005597210.1 | 1662 | 63 | |||
XM_014738562.2 | XP_014594048.1 | 1626 | 63 | |||
XM_023651999.1 | XP_023507767.1 | 1662 | 63 | |||
XM_023652000.1 | XP_023507768.1 | 2124 | 63 | |||
XM_023652001.1 | XP_023507769.1 | 2439 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
