Assay Details
Gene Symbol: SLC25A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495909.4 | XP_001495959.1 | 886 | 61 | ||
XM_005597165.3 | XP_005597222.1 | 1059 | 61 | |||
XM_005597166.3 | XP_005597223.1 | 972 | 61 | |||
XM_014738702.2 | XP_014594188.1 | 957 | 61 | |||
XM_023652024.1 | XP_023507792.1 | 1145 | 61 | |||
XM_023652025.1 | XP_023507793.1 | 1006 | 61 | |||
XM_023652026.1 | XP_023507794.1 | 1081 | 61 | |||
XM_023652027.1 | XP_023507795.1 | 822 | 61 | |||
XM_023652028.1 | XP_023507796.1 | 1223 | 61 | |||
XM_023652029.1 | XP_023507797.1 | 1014 | 61 | |||
XM_023652030.1 | XP_023507798.1 | 995 | 61 | |||
XM_023652031.1 | XP_023507799.1 | 908 | 61 | |||
XM_023652032.1 | XP_023507800.1 | 963 | 61 | |||
XM_023652033.1 | XP_023507801.1 | 1050 | 61 | |||
XM_023652034.1 | XP_023507802.1 | 757 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
