Assay Details
Gene Symbol: MIF4GD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738743.2 | XP_014594229.1 | 402 | 100 | ||
XM_023652041.1 | XP_023507809.1 | 553 | 100 | |||
XM_023652043.1 | XP_023507811.1 | 402 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
