Assay Details
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496574.4 | XP_001496624.1 | 497 | 69 | ||
XM_014738776.2 | XP_014594262.1 | 415 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
