Assay Details
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652074.1 | XP_023507842.1 | 1208 | 69 | ||
XM_023652075.1 | XP_023507843.1 | 1155 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
