Assay Details
Gene Symbol: GPRC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738985.2 | XP_014594471.1 | 1207 | 74 | ||
XM_014738990.2 | XP_014594476.1 | 1174 | 74 | |||
XM_023652095.1 | XP_023507863.1 | 1079 | 74 | |||
XM_023652096.1 | XP_023507864.1 | 1430 | 74 | |||
XM_023652097.1 | XP_023507865.1 | 1330 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
