Assay Details
Gene Symbol: RPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362519.4 | XP_003362567.2 | 297 | 74 | ||
XM_005597217.3 | XP_005597274.1 | 136 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
