Assay Details
Gene Symbol: TACO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501153.4 | XP_001501203.1 | 815 | 83 | ||
XM_005597286.3 | XP_005597343.1 | 870 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
