Assay Details
Gene Symbol: GOSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001487945.5 | XP_001487995.1 | 307 | 69 | ||
XM_005597305.3 | XP_005597362.1 | 307 | 69 | |||
XM_014739593.2 | XP_014595079.1 | 205 | 69 | |||
XM_023652346.1 | XP_023508114.1 | 307 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
