Assay Details
Gene Symbol: RDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917213.4 | XP_001917248.1 | 784 | 66 | ||
XM_023652350.1 | XP_023508118.1 | 1047 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
