Assay Details
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652403.1 | XP_023508171.1 | 807 | 86 | ||
XM_023652404.1 | XP_023508172.1 | 720 | 86 | |||
XM_023652405.1 | XP_023508173.1 | 716 | 86 | |||
XM_023652406.1 | XP_023508174.1 | 689 | 86 | |||
XM_023652407.1 | XP_023508175.1 | 724 | 86 | |||
XM_023652408.1 | XP_023508176.1 | 1642 | 86 | |||
XM_023652409.1 | XP_023508177.1 | 734 | 86 | |||
XM_023652410.1 | XP_023508178.1 | 832 | 86 | |||
XM_023652411.1 | XP_023508179.1 | 582 | 86 | |||
XM_023652412.1 | XP_023508180.1 | 629 | 86 | |||
XM_023652413.1 | XP_023508181.1 | 522 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
