Assay Details
Gene Symbol: PPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495667.4 | XP_001495717.2 | 71 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001495667.4 transcripts.
Set Membership: |
![]() ![]() |
