Assay Details
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494084.6 | XP_001494134.1 | 587 | 68 | ||
XM_001494111.5 | XP_001494161.1 | 733 | 68 | |||
XM_001494140.6 | XP_001494190.1 | 498 | 68 | |||
XM_005597373.3 | XP_005597430.1 | 588 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
