Assay Details
Gene Symbol: RAB5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494956.4 | XP_001495006.1 | 691 | 60 | ||
XM_023652516.1 | XP_023508284.1 | 553 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
