Assay Details
Gene Symbol: GSDMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362385.4 | XP_003362433.2 | 1561 | 64 | ||
XM_023652578.1 | XP_023508346.1 | 2808 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
