Assay Details
Gene Symbol: ZPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740694.2 | XP_014596180.1 | 623 | 78 | ||
XM_023652583.1 | XP_023508351.1 | 956 | 78 | |||
XM_023652584.1 | XP_023508352.1 | 957 | 78 | |||
XM_023652585.1 | XP_023508353.1 | 788 | 78 | |||
XM_023652586.1 | XP_023508354.1 | 663 | 78 | |||
XM_023652587.1 | XP_023508355.1 | 635 | 78 | |||
XM_023652588.1 | XP_023508356.1 | 566 | 78 | |||
XM_023652589.1 | XP_023508357.1 | 550 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
