Assay Details
Gene Symbol: LASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740826.2 | XP_014596312.1 | 368 | 80 | ||
XM_023652626.1 | XP_023508394.1 | 465 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
