Assay Details
Gene Symbol: SCRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501869.5 | XP_001501919.1 | 640 | 60 | ||
XM_023652658.1 | XP_023508426.1 | 734 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
