Assay Details
Gene Symbol: CBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502003.6 | XP_001502053.1 | 200 | 71 | ||
XM_023652674.1 | XP_023508442.1 | 382 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
