Assay Details
Gene Symbol: PCTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500249.5 | XP_001500299.3 | 736 | 94 | ||
XM_023652824.1 | XP_023508592.1 | 793 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
