Assay Details
Gene Symbol: DYNLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597508.3 | XP_005597565.1 | 209 | 95 | ||
XM_023652866.1 | XP_023508634.1 | 457 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
