Assay Details
Gene Symbol: C11H17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504001.5 | XP_001504051.1 | 499 | 81 | ||
XM_005597587.3 | XP_005597644.1 | 492 | 81 | |||
XM_014741925.2 | XP_014597411.2 | 942 | 81 |
