Assay Details
Gene Symbol: C11H17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504001.5 | XP_001504051.1 | 848 | 119 | ||
XM_005597587.3 | XP_005597644.1 | 841 | 119 | |||
XM_014741925.2 | XP_014597411.2 | 1291 | 119 |

More Information
Set Membership: |
![]() ![]() |
